Would you like to log in again?
Funnel, High Density Polyethylene, Dynalon
Supplier: Dynalon
212105 212145 212115
71070-014CS 168.55 USD InStock
71070-014 71070-020 71070-012
Funnel, High Density Polyethylene, Dynalon
Funnels Filtration Funnels Filter Funnels
Funnels are ideal for many applications in the laboratory and industrial markets
Upper portion of funnel is straight sided to helps prevent spill-over. Maximum temperature for HDPE is 110°C.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty