Would you like to log in again?
Tubing Adapters, Polypropylene, Dynalon
Supplier: Dynalon
227565 227545
71070-028CS 520.8 USD InStock
71070-028 71070-032
Tubing Adapters, Polypropylene, Dynalon
Fittings and Connectors
Adapters allow for connecting different diameter tubing.
  • Excellent chemical resistance
  • Serrated surfaces to prevent slippage
  • Autoclavable
Features serrated connecting surfaces to prevent tubing slipping. Made of autoclavable polypropylene with excellent chemical resistance.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty