Would you like to log in again?
Stirrer Blade, Polypropylene, Dynalon
Supplier: Dynalon
212615
71070-132CS 432.45 USD InStock
71070-132
Stirrer Blade, Polypropylene, Dynalon
Stirrers Stirring Blades
Centrifugal polypropylene stirrer blades are 100mm in width (fully extended)
The blades are made of chemically resistant polypropylene and are fixed to a PVC-covered steel steel rod with a diameter of 8x350mm (approx. ⁵⁄₁₆ x 13¾") long. Clockwise motion directs flow upward.
Ordering information: Elasticity of tubing will influence choice of size. Dimensions listed are nominal.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty