Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Round Dipping Baskets, Dynalon
Supplier: Dynalon
146604 146644 146594
71070-280EA 526.51 USD InStock
71070-280 71070-274 71070-242
Round Dipping Baskets, Dynalon
Baskets Safety Transporting Baskets
Round dipping baskets are fabricated of either 1/8” thick HDPE or PP
HDE takes 140°F on a continuous basis or more for shorter periods (up to 160°F). Polypropylene fabricated dipping baskets withstands 180°F on a continuous basis or up to 200°F for shorter periods. Both are supplied with sturdy swinging handle—a unique feature allowing easy storage and loading. All have perforated bottoms. Excellent chemical resistance.
Ordering information: Additional sizes and materials are available upon request.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty