Would you like to log in again?
ADAPTER F/EBERBACH SHAKRS
Supplier: EBERBACH CORP
Adapter^ To increase shaking capacity. For use with reciprocating Eberbach* shakers, Models 6000 and 6500.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
ADAPTER F/EBERBACH SHAKRS 71470-034 E6050.00 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty