Would you like to log in again?
Citrus Clearing Solvent, Epredia
Supplier: Epredia
8301 8301
72060-044CS 974.03 USD InStock
72060-044 CA72060-044
Citrus Clearing Solvent, Epredia
Stains Microscopy and staining reagents
Replace xylene in both manual and automated system applications with Epredia™ Citrus Clearing Solvent.
  • D-limonene-based solvent that has a slight citrus scent
  • Higher flash point eliminates the need for storage in a flammable safety cabinet
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty