Would you like to log in again?
Petroff-Hausser Counting Chamber, Hausser Scientific
Supplier: Hausser Scientific
3900 3920
15170-048EA 908.01 USD InStock
15170-048 72350-004
Petroff-Hausser Counting Chamber, Hausser Scientific
Slides Counting Chambers
Designed specifically for counting bacteria, sperm, blood platelets, and the content of vaccines
The 0.02mm cell depth minimizes the necessity of focusing up and down to identify organisms. Housing protects chamber from scratching during use. Suitable size for use in dark field microscopy. Improved Neubauer ruling pattern etched permanently onto the polished surface. One piece construction.
Ordering information: The Petroff-Hausser Counting Chamber is supplied with 2 cover glasses.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty