Would you like to log in again?
Kimble® Disposable Culture Tubes, Round Bottom, Screw-Cap Finish, DWK Life Sciences
Supplier: DWK Life Sciences
90K16100F 73770-13100 73770-16150 73770-1690 73770-16100 73760-16125 73770-20150 73770-16125 73770-20125
72690-018CS 425.65 USD InStock
72690-018 76212-366 72690-016 60825-837 72690-028 72690-022 72690-024 CA60826-153XFB 72690-020
Kimble® Disposable Culture Tubes, Round Bottom, Screw-Cap Finish, DWK Life Sciences
Tubes Culture Tubes
Tubes are designed for use in tissue culture work and general bacteriological applications.
  • Supplied without a marking area and without caps
  • Tubes are shrink-wrapped in plastic film in modular trays
  • Resistant to chemical, thermal and mechanical shock
  • Dishwasher safe and patented inner clamps keep tubes centered and vertical
Tubes are manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements.
Please note: The "capacity" given below indicates the brim (overflow) capacity.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty