Would you like to log in again?
BioRobotix® Pipette Tips for MultiMek™ Workstations, Molecular BioProducts
Supplier: Molecular BioProducts
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
BioRobotix®
918-021 919-262 918-262 919-021 919-261
72830-230CS 785.89 USD InStock
72830-230 72830-220 72830-228 72830-224 72830-226
BioRobotix® Pipette Tips for MultiMek™ Workstations, Molecular BioProducts
Pipette Tips
Compatible with MultiMek™ style workstations/AP96.
  • Sterile tips are certified RNase-, DNase-, DNA-, and pyrogen-free
Available as clear polypropylene only, in 20, 130 and 250 µl sizes. Filter tips utilize the ART barrier for guranteed protection against cross-contamination.
Ordering information: Tips are packaged 96 per tray, 10 trays per pack.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty