Would you like to log in again?
TIP FLTR AP96 CLEAR 20UL PK960
Supplier: Thermo Scientific
MBP BioRobotix* MultiMek* Style Tips^ 20uL working volume. Racked, sterile. Designed with clear polypropylene for AP96 workstation. RNase-, DNase-, DNA-, and pyrogen-free.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TIP FLTR AP96 CLEAR 20UL PK960 72830-222 918-261 Case of 4800 Retrieving
Pack of 960 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty