Would you like to log in again?
BioRobotix® Pipette Tips for Rosys® and QIAGEN® Workstations, Molecular BioProducts
Supplier: Molecular BioProducts
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
BioRobotix®
953-252 953-261 951-251 953-262 953-011 951-011 953-251 951-261 953-021
72830-252CS 214 USD InStock
72830-252 72830-264 72830-266 72830-244 72830-260 72830-262 72830-256 72830-258 72830-248
BioRobotix® Pipette Tips for Rosys® and QIAGEN® Workstations, Molecular BioProducts
Pipette Tips
Compatible with Rosys® and QIAGEN® style workstations.
  • Sterile tips are certified RNase-, DNase-, DNA-, and pyrogen-free
Available as liquid sensing tips to maximize the workstation abilites, or clear polypropylene for applications that do not require liquid sensing. Three volume sizes include 250, 300 and 1100 µl. Filter tips utilize the ART® barrier.
Ordering information: Tips are packaged 96 per tray, 10 trays per case.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty