Would you like to log in again?
Automation Pipette Tips for Qiagen BioRobot Workstations, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
951-253 953-253
72830-334CS 1053.34 USD InStock
72830-334 72830-336
Automation Pipette Tips for Qiagen BioRobot Workstations, Thermo Scientific
Pipette Tips
Thermo Scientific provides a solution for your Qiagen BopRobot pipette tips.
These automation pipette tips have been designed, validated, and tested on a Qiagen BioRobot to ensure equal performance without the need to modify your existing protocols and programs. Our low percentage Coefficient of Variation (%CV) ensures product consistency and performance from every lot. Each lot must pass a 15-point QC inspection including performance testing on our BioRobot workstation before tips are released. All tips are certified RNase, DNase, DNA, endotoxin, bioburden, and pyrogen free.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty