Would you like to log in again?
HydroLogix HLT Blue Pipette Tips, Molecular BioProducts
Supplier: Molecular BioProducts
3950-11 3951-11
72830-684CS 782.88 USD InStock
72830-684 72830-688
HydroLogix HLT Blue Pipette Tips, Molecular BioProducts
Pipette Tips
These non-filtered pipette tips utilizes a unique bi-fabricated wall that has been designed to produce a secure seal at a fraction of the force required to load standard pipette tips onto the pipettor. Accordingly, the forces required to eject the tips from your pipettor are significantly reduced.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty