Would you like to log in again?
Diff-Safe® Blood Dispensers, Alpha Scientific
Supplier: Alpha Scientific
Diff-Safe®
125 BULK 101 101
73000-075CS 7102.79 USD InStock
CA73000-077 73000-075 73000-077
Diff-Safe® Blood Dispensers, Alpha Scientific
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
Designed to minimize airborne pathogens by eliminating the need to remove the stopper from the blood tube
The dispensers feature a cannula that inserts easily through the rubber stopper of a blood collection tube. When the tube is inverted and the dispenser pressed against a slide, it dispenses a drop ideally sized for making a blood smear. Compatible with BD Vacutainer® tubes with BD Hemogard™ closures. Nonsterile.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty