Would you like to log in again?
VWR® qPCR Master Mix
Supplier: Avantor
VWR®
M3021B-AV1 VWRC73112-0500 M3021B-AV2 VWRC73112-0500
73112-1000EA 474.96 USD InStock
73112-1000 73112-2500 73112-0500 73112-0100
VWR® qPCR Master Mix
Nucleic Acid Reagents qPCR/RT-qPCR Enzymes and Kits
VWR® qPCR Master Mix features a distinctive universal passive reference dye that is compatible with ROX-dependent and ROX-independent qPCR instruments.
  • For the detection and quantification of target DNA sequences using the SYBR®/FAM channel
  • Ready-to-use master mix containing all necessary components for DNA amplification and quantification, except for primers and DNA template
  • Suitable for both genomic DNA and cDNA
  • Includes Hot Start Taq DNA Polymerase for optimal performance
  • Incorporates dUTP for carryover contamination prevention
  • Supplied at a 2X concentration
VWR® qPCR Master Mix is designed to facilitate rapid, sensitive, and precise detection and quantification of specific DNA and cDNA sequences via dye-based qPCR.
Caution: Store at -20°C. For research use only.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty