Would you like to log in again?
VWR® One-Step RT-qPCR Kit
Supplier: Avantor
VWR®
VWRC73212-0500 E3023B-AV2 VWRC73212-0500 E3023B-AV1
73212-2500EA 2141.52 USD InStock
73212-2500 73212-0500 73212-1000 73212-0100
VWR® One-Step RT-qPCR Kit
Nucleic Acid Reagents qPCR/RT-qPCR Enzymes and Kits
VWR® One-Step RT-qPCR Kit is designed to facilitate rapid, sensitive, and precise detection and quantification of various RNA sequences via dye-based qPCR. It features a distinctive universal passive reference dye that is compatible with ROX-dependent and ROX-independent qPCR instruments.
  • For the detection and quantification of RNA targets using the SYBR®/FAM channel
  • Ready-to-use kit containing all necessary reaction components, except for primers and RNA template
  • Suitable for a variety of RNA sample types
  • Includes Hot Start Taq DNA Polymerase and VWR® Reverse Transcriptase for improved reaction specificity
  • Incorporates dUTP for carryover contamination prevention
  • VWR® One-Step Reaction Mix is supplied at a 2X concentration
The VWR® RT Enzyme Mix, supplied at a 20X concentration, contains the specially designed, temperature-controlled VWR® Reverse Transcriptase and Murine RNase Inhibitor to assist in preventing RNA degradation. This temperature-controlled RT enzyme prevents unwanted non-specific activity and displays superior thermostability in comparison to other reverse transcriptases.
Caution: Store at -20°C. For research use only.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty