Would you like to log in again?
Nalgene® Diagnostic Bottles, PETG, Sterile, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
342035-0005 342035-0020 342035-0010 2035-0010 2035-0020 2035-0005
73520-082CS 265.92 USD InStock
73520-082 73520-084 73520-080 73520-086 73520-078 73520-088
Nalgene® Diagnostic Bottles, PETG, Sterile, Thermo Scientific
Bottles Plastic Bottles
PETG single-use diagnostic bottles are ideal for sampling, storage, and shipment of reagents and buffer solutions.
  • Bottles and caps are gamma sterilized to 10⁻⁶ sterility assurance levels and have a lot number
  • Nonpyrogenic, noncytotoxic
Ordering information: Supplied with white, lined screw caps with a 20-415 thread finish. Tray-packed bottles are packed 20 bottles per tray, 5 trays per case. Bulk-packed bottles are packed 100 bottles per tray, 5 trays per case. All trays are packaged in double-lined cartons for cleanroom use, with caps assembled.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty