Would you like to log in again?
Nalgene® Serum Vial Closures, HDPE, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Nalgene®
342158-0021 312158-0021 342158-0023 342158-0025 342158-0024 342158-0026
73520-094CS 1048.32 USD InStock
73520-094 73520-096 73520-090 73520-092 16059-992 16060-100
Nalgene® Serum Vial Closures, HDPE, Thermo Scientific
Closures Chromatography Vial Closures
These 20mm, high-density polyethylene screw cap closures are available lined or unlined and sterile or nonsterile.
Ordering information: All closures are bulk packed. Sterile closures are bulk packed in a case with a three-ply liner. Unlined closures require a stopper (not included).
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty