Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nalgene® Safety Waste Systems, Thermo Scientific

Supplier: Thermo Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Nalgene®
6379-0010 6379-0004
73520-204CS 321.45 USD InStock
73520-204 73520-202
Nalgene® Safety Waste Systems, Thermo Scientific
Bottles Solution Bottles
For temporary disposal of most chemical and biological wastes. Each system consists of a waste container, removable HDPE funnel and cover, polypropylene closure/funnel adapter, and PTFE vent membrane.

Red, hinged cover seals funnel, reducing the level of volatiles evaporating into the air. Easy-snap safety latch minimizes spillage if system is tipped over. Redesigned vent system with filter reduces fumes and splashing. Large-diameter funnels, in addition to the vents, enable fast and easy pouring. Funnel stem extends below the liquid level to reduce the evaporation rate. Removable neck screen helps trap large particles, like stir bars.

Plastic containers are durable, chemical-resistant and break-resistant — safer than glass. Safety waste funnels and 4L (1.1gal.) waste containers are made of high-density polyethylene. The 10L (2.6gal.) waste container is made of fluorinated high-density polyethylene.

Note: For temporary waste disposal only. Bottles are not safety cans as defined by OSHA guidelines.

Closed system makes waste disposal safer and easier.

Ordering information: The 4L (1.1gal.) system includes safety waste funnel 73520-198 and a 4L (1.1gal.) narrow mouth bottle. The 10L (2.6gal.) system includes safety waste funnel 73520-200 and a 10L (2.6gal.) fluorinated carboy. Secondary containers (sold separately) should be used with Safety Waste Systems during transport, transfer or storage; contact your Avantor representative more more information.

Order Now
 

ORDER

ACCESSORIES

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty