Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Nalgene® Disposable Polypropylene Robotic Reservoirs, Thermo Scientific
Supplier: Thermo Scientific
1200-2301 1200-1301 1200-2300 1200-1300
73520-392CS 422.37 USD InStock
73520-392 73520-390 73520-388 73520-386
Nalgene® Disposable Polypropylene Robotic Reservoirs, Thermo Scientific
Reagent Reservoirs
Nalgene Disposable Polypropylene Robotic Reservoirs come with either a convoluted bottom design or a flat bottom design.
  • Convoluted design minimizes dead volume and has baffles to reduce splashing
  • Convoluted bottom design is most effective with a 96-well format
  • Flat Bottom design can be used with single, 6-, 12-, 96-, 384- and 1536-well formats
These reservoirs are compatible with most robotic arms, plate stackers, plate cranes, and platform configurations.
Certifications: Meets the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty