Would you like to log in again?
Nunc® InVitro Roller Bottles, PETG, Sterile, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Nunc®
2160-20 1060-85 2160-05 4260-22 2160-05 1760-20 1060-85 2160-20 1060-20 1860-22 1760-20 1060-05 1060-05 1060-20 1860-22 4260-22
73521-060CS 333.93 USD InStock
73521-060 CA73520-418 CA73520-734 73520-418 CA73521-060 73520-734 CA73520-732 CA44200-012 73520-732 73520-414 CA62404-388 CA73520-420 CA73520-410 73520-412 73520-420 73520-410
Nunc® InVitro Roller Bottles, PETG, Sterile, Thermo Scientific
Bottles Roller Bottles
Tough, flexible PETG roller bottles offer a safer, more convenient alternative to conventional polystyrene roller bottles.
  • Bottles include a 48 mm standard cap, except for the bottles with a vented cap
  • Printed, easy-to-read graduations aid in the measurement of medium fills
Standard surface bottles offer 1050 cm² of surface area, providing more capacity than 850 cm² bottles for adherent or suspension cultures. They can also be used as freezing containers in temperatures as low as –40 °C (–40 °F).
Expanded surface roller bottles have specially-designed pleats that expand the available growth surface, providing up to 2.5 times the growth area of flat-wall bottles. Pleats are oriented in the direction of rotation to minimize turbulence while rolling. Flat panels between sections of pleats allow microscopic viewing and drainage during medium replacement and harvest.
Bottles are blown-molded and have no sonic welds or other leak-prone joints. Lot number and expiration date are printed on each bottle to maximize traceability.
Ordering information: Sterile packed in bulk packaging or in lab packs of five bottles. Vented caps are sterile and individually wrapped within a bulk pack.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty