Would you like to log in again?
Nalgene® High Profile Closures with Color Coders for Micro Packaging Vials, Thermo Scientific
Supplier: Thermo Scientific
362830-5110 362830-0119 362830-0113 362830-0111 362830-0118 362830-0112 362830-0110 362830-0117 362830-5116 362830-5118 362830-0116 362830-5114 362830-0114 362830-0115
73520-816CS 341.04 USD InStock
73520-816 73520-814 73520-438 73520-812 73520-820 73520-436 73520-810 73520-808 73520-806 73521-330 73520-818 73521-328 73520-434 73520-432
Nalgene® High Profile Closures with Color Coders for Micro Packaging Vials, Thermo Scientific
Closures Test/Sample Tube Closures
Nalgene Closures with Color Coders are made of high density polyethylene, and have no O-ring to fall out or contaminate contents
The color coders are permanently attached, are free of toxic heavy metals, and don not come in contact with vial contents. These high profile closures are non-cytotoxic and non-pyrogenic.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty