Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Nalgene® Rapid-Flow™ Filter Units and Bottle Top Filters, PES Membrane, Sterile, Thermo Scientific
Supplier: Thermo Scientific
Nalgene® MF75™
597-4520 564-0020 295-4545 596-3320 166-0045 567-0020 595-3320 569-0020 165-0045 295-3345 566-0020 168-0045 296-4545 568-0010 596-4520 567-0010 568-0020 595-4520 169-0045 566-0010 565-0020 167-0045 597-3320 296-3345 565-0010
89176-982CS 386.53 USD InStock
89176-982 16211-066 73520-988 16211-068 73520-986 73520-994 97014-834 73520-992 16211-064 73520-990 16211-061 73520-998 16211-056 16211-077 97014-830 73520-996 97014-832 73521-002 73521-000 73520-984 16211-073 16211-054 82030-938 16211-053 73520-982
Nalgene® Rapid-Flow™ Filter Units and Bottle Top Filters, PES Membrane, Sterile, Thermo Scientific
Filters Bottle Top Filters
All of these filters come with an asymmetric polyethersulfone (PES) membranes.
  • Graduated upper chambers hold from 150 to 1000 ml
  • Asymmetric PES membrane is an extremely clean, low-extractable, fast-flowing membrane
  • Provides excellent throughput and low protein binding of media and other solutions
  • 0.2 µm pore size membrane provides a last line of defense against bacterial contamination
  • 0.1 µm pore size membranes also protect against mycoplasma contamination
  • 0.45 µm pore size membranes are suitable for particle analysis, scintillation counting, prefiltration of aqueous solutions, and general cytology
Complete units come with a bottle-top filter and a receiver storage bottle. Bottle-top filters have side arms fitted with quick-disconnect tubing adapters and screw securely into 33 or 45 mm neck size bottles.
All filters are printed with catalog number, membrane material, pore size, lot number, and expiration date.
Certifications: Certified radiation-sterilized, non-cytotoxic, and non-pyrogenic.
Caution: Nalgene® Rapid-Flow™ Filter Units and Bottle Top Filters are designed for laboratory use only, not for in vitro diagnosis or parenterals.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty