Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Nunclon™ Delta MultiDishes, Sterile, Thermo Scientific
Supplier: Thermo Scientific
Nunclon™
176740 140685 140675 150687 142475 150628 142485 140675 142485 152640 152640 176740 140685 150687 150628 142475
73520-836CS 361.7 USD InStock
CA73520-836 73520-836 73521-004 CA73520-906 73520-906 CA73521-008 CA62407-068 CA62407-332 73521-006 CA73521-004 CA73521-006 73521-008 62407-068 62407-332 62407-338 CA62407-338
Nunclon™ Delta MultiDishes, Sterile, Thermo Scientific
Culture Dishes Cell Culture Dishes
These polystyrene dishes with flat-bottom wells incorporate the Nunclon™Δ treated surface for optimal cell attachment and growth
Lids are keyed for a one-way fit. Rings in the lids reduce evaporation and condensation. Writing areas on both lids and dishes and individual alphanumeric markings allow easy identification of samples. Nontoxic and certified nonpyrogenic. Stackable.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty