Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Nunc® Coated MultiDishes, Thermo Scientific
Supplier: Thermo Scientific
Nunc®
152035 152034 152034 152025 152035
76331-200CS 378.09 USD InStock
CA73521-010 76331-200 73521-024 CA73521-024 73521-010
Nunc® Coated MultiDishes, Thermo Scientific
Culture Dishes Cell Culture Dishes
Enhance cell attachment to the culture surface with the extracellular matrix coating on Nunc™ Poly-D-Lysine or Collagen I Coated Multidishes.
  • Aseptically sterile and non-pyrogenic
  • Poly-D-Lysine is a chemically synthesized animal-free material
  • Collagen I is of animal origin
Ideal for culturing finicky cells that do not attach to the regular cell culture-treated surfaces.
Ordering information: Dishes come packed in sleeves of five.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty