Would you like to log in again?
Nalgene® Graduated Square Bottles, PC, Tray Packed, without Closures, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
362015-0500 362015-1000 362015-0125 362015-0250
73521-082CS 195.84 USD InStock
73521-082 73521-080 73521-078 73521-076
Nalgene® Graduated Square Bottles, PC, Tray Packed, without Closures, Thermo Scientific
Bottles Plastic Bottles
Nalgene® graduated square bottles are polycarbonate, transparent, and autoclavable.
They have an easy-to-handle square design and molded-in graduations with a drip-proof neck. These bottles are ideal for storage of aqueous solutions and short-term storage of media.
Ordering information: These bottles are packaged in bulk. The number of bottles in the package depends upon the size of the bottle.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty