Would you like to log in again?
Nunc® Immobilizer™ Amino Plates and Modules, Thermo Scientific
Supplier: Thermo Scientific
Nunc® Immobilizer™
436008 436008 436006 436013 436023 436013 436007 436023 436006 436007
73521-126CS 915.57 USD InStock
CA73521-128 CA73521-256 CA73521-124 73521-126 CA73521-126 73521-128 73521-254 73521-256 CA73521-254 73521-124
Nunc® Immobilizer™ Amino Plates and Modules, Thermo Scientific
Microplates Immunology Microplates and Strips
These plates are ideal for one-step covalent coupling of peptides, proteins, and aminated oligos with minimal denaturation
Clear plates guarantee uniform well-to-well binding capacity of CV ≤5%; black and white plates guarantee uniform well-to-well binding capacity of CV ≤7.5%.
Free amino or subhydryl groups on the biomolecule react spontaneously and are covalently tethered to the surface through an optimized spacer arm. Choose from polystyrene plates in standard 96-well format or polystyrene modules consisting of twelve 8-well strips. LockWell™ modules feature 8-well break apart strips that can be separated into individual wells and then locked into a frame at the same horizontal level for uniform washing and reading. Plates have a high signal-to-noise ratio, no surface leaching, reduced steric hindrance of bound molecules, and highly reproducible binding with low CVs. They require no blocking or activation steps and feature site directed binding. Plates are also stable for long periods at room temperature. Choose clear plates and strips for colorimetric assays, white plates for bioluminescence and chemiluminescence assays, and black plates for fluorescence assays.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty