Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Nunc® Multidishes, Non-treated, Thermo Scientific
Supplier: Thermo Scientific
144530 150200 144530 179820 150787 179820 150787 150200 150239 150239
73521-138CS 287.83 USD InStock
CA73521-356 73521-138 CA73521-138 CA73521-142 73521-144 CA73521-144 73521-356 73521-140 73521-142 CA73521-140
Nunc® Multidishes, Non-treated, Thermo Scientific
Culture Dishes Cell Culture Dishes
These multi dishes are non-treated and can be used for the suspension of cell cultures
The dishes have raised well rims which lowers the risk of cross-contamination. They also have flat bottom wells that allow for easy viewing of cultures. Lids with notched corners for a good fit are included. The lids help with single position orientation and cross contamination prevention. The multi dishes are made of sterile polystyrene.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty