Would you like to log in again?
Nalgene® HDPE High Profile Closures with Color Coders for Micro Packaging Vials, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
342830-0119 342830-5114 342830-0110 342830-5116 342830-0116 342830-5118 342830-0115 342830-0114 342830-0112 342830-0113 342830-1110 342830-0111 342830-0118 342830-5110
73521-180CS 484.33 USD InStock
73521-180 73521-170 73521-182 73521-172 73521-190 73521-188 73521-166 73521-178 73521-168 73521-162 73521-184 73521-174 73521-186 73521-164
Nalgene® HDPE High Profile Closures with Color Coders for Micro Packaging Vials, Thermo Scientific
Closures Test/Sample Tube Closures
The Thermo Scientific Nalgene HDPE closures for micro packaging vials have no o-ring to fall out or contaminate contents
These closures are provided sterile and are for use with our sterile micro packaging vials. The closures are also leakproof, non-cytotoxic, and non-pyrogenic.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty