Would you like to log in again?
Nunc™ Cell Culture Flasks, Thermo Scientific
Supplier: Thermo Scientific
Nunc® EasYFlask™
156340 132708 156472 156800 178883 156499 169900 159910 153732 163371 132097 163371 132703 132705 169900 132707 178883 159934 156499 178885 132706 132705 156340 132703 136196 136196 156472 132706 159934 159920 132704 178983 132097 132098 156502 156502 153732 178985 159926 156367 178905 178905 132704 156367 156800 159933 159926 156505 159933 178983 132098 132708 159910 159920 132707
15708-124CS 461.99 USD InStock
15708-124 10000-968 CA15708-134 30617-430 15708-094 15708-096 15708-097 15708-130 CA30617-430 15708-134 CA89020-034 CA89020-032 CA15708-101 CA15708-103 CA89020-038 CA15708-106 CA89020-036 30617-428 73521-146 CA73521-150 CA73521-146 CA73521-148 89020-036 15708-101 CA89020-042 15708-103 89020-034 73521-150 15708-106 CA12777-958 89020-038 10000-982 10000-980 CA30617-428 CA73521-200 CA73521-360 73521-148 CA73521-358 CA12777-960 CA89020-040 89020-032 CA15708-120 CA15708-124 12777-958 73521-200 73521-360 73521-358 CA15708-094 CA15708-096 15708-120 89020-042 12777-960 89020-040 CA15708-097 CA15708-130
Nunc™ Cell Culture Flasks, Thermo Scientific
Flasks Cell Culture Flasks
These Nunc™ cell culture flasks are designed for culture consistency, cell health, and reproducibility. Choose from a variety of surfaces and sizes with culture areas ranging from 25 to 500 cm² to suit your specific applications and cell types.
  • Nunclon Delta certification including testing on four different cell types is provided in each case
  • Barcoded 175 cm² flasks (*) with large Code 128 are compatible with automation
  • Sterilized by gamma irradiation and certified to be non-pyrogenic
Flasks treated with the proprietary Nunclon™ Delta surface treatment ensure consistent cell growth and maximum adhesion for a broad range of cell types. Nunc™ non-treated flasks are made with high-quality, optically clear virgin polystyrene with a hydrophobic surface ideal for suspension cell culture and useful for a variety of biochemical assays.
The ergonomic design of Nunc™ EasYFlask with 1/3-turn cap enables one-handed operation and avoids wrist strain. The angled, extra-wide neck provides easier access to growth surface with cell scrapers or pipettes. Molded and printed graduations help enable easy and quick measurement of growth media. Nunc™ standard flasks are designed with a straight neck and barcoding option for automation cell culture.
Options of solid or filtered cap for adequate gas exchange. Solid caps are vent/close caps with 'Y' marks that allow easy visual verification of venting position while promoting sufficient gas exchange. Filter caps ensure constant gas exchange and constant airflow while minimizes chances of contamination.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty