Would you like to log in again?
Nunc CryoTube Colored Caps, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
336092 336088 363436 336094 336081 363436 336094 336092 336081
73521-221CS 1102.8 USD InStock
CA12020-008 CA12020-006 73521-221 73521-222 73521-223 73521-394 CA12020-002 CA73521-394 73521-220
Nunc CryoTube Colored Caps, Thermo Scientific
Closures Test/Sample Tube Closures
Nunc CryoTube colored caps are replacement polypropylene screw caps with a silicone gasket to provide the best seal
They are made to be used with Nunc internally threaded CryoTube vials.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty