Would you like to log in again?
Nalgene® Graduated Square Bottles, PPCO, Bulk Pack, Thermo Scientific
Supplier: Thermo Scientific
312016-0060 312016-0500 312016-1000 312016-0125
73521-287CS 666.67 USD InStock
73521-287 46610-188 73521-284 16059-484
Nalgene® Graduated Square Bottles, PPCO, Bulk Pack, Thermo Scientific
Bottles Plastic Bottles
Nalgene® graduated polypropylene copolymer square bottles are autoclavable, and have molded-in graduations for easy measuring.
The bottles feature a square design which helps save space. These bottles have a drip-proof neck finish for convenient dispensing.
Ordering information: Bottles are packaged in bulk. The number of bottles in the package depends on the size of the bottle.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty