Would you like to log in again?
Nalgene® Boston Bottles, Round, without Closures, HDPE, Bulk Pack, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
362002-0001 362002-0016 362002-0002 362002-0008 362002-0004
16211-000CS 934.08 USD InStock
16211-000 73521-314 73521-316 16211-002 73521-312
Nalgene® Boston Bottles, Round, without Closures, HDPE, Bulk Pack, Thermo Scientific
Bottles Plastic Bottles
Nalgene® high density polyethylene Boston Round Bottles are translucent and have excellent chemical resistance.
These bottles have durable uniform walls that are resistant to splitting or puncturing. They are specifically designed for demanding packaging applications.
Certifications: Manufactured from validated and controlled resins meeting a variety of food and pharmaceutical regulatory requirements.
Ordering information: Bottles are bulk packed for packaging applications. For closures, see 16060-016 series.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty