Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Nunc™ Petri Dishes, Thermo Scientific
Supplier: Thermo Scientific
240835 249964 263991 249964 4021 263991
25387-024CS 1121.98 USD InStock
25387-024 73521-370 73521-368 CA73521-368 25384-002 CA73521-370
Nunc™ Petri Dishes, Thermo Scientific
Culture Dishes Petri Dishes
Nunc™ Petri Dishes are available in a variety of styles for laboratory use or for use with automatic filling equipment. Most dishes are vented to permit gas exchange during incubation. Bottom flatness is carefully controlled to ensure even distribution of media and organisms. The Bio-Assay dish was developed for use in agar diffusion assays (e.g., antibiotic activity determinations).
  • Large square area is especially useful for screening libraries and isolating clones
  • Dishes are suitable for culturing of fungi, bacteria and other microorganisms
  • Compatible with automated robotic instrumentation
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty