Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Nunc® Rectangular Dishes, Polystyrene, Sterile, Thermo Scientific
Supplier: Thermo Scientific
Nunc®
267062 267062 267060 267060 267061
73521-428CS 609.66 USD InStock
73521-428 CA73521-428 73521-424 73521-420 CA73521-420
Nunc® Rectangular Dishes, Polystyrene, Sterile, Thermo Scientific
Culture Dishes Petri Dishes
These 1-, 4-, and 8-well rectangular dishes feature an untreated surface for molecular biology applications
The 1-well dish is ideal for HTS petri dish applications. The 4- and 8-well dishes are ideal for compartmentalizing assays, probing multiple gene subsets, and washing Western blot strips. The 4-well dish can hold four standard glass slides. All dishes are made of polystyrene and feature ANSI standard dimensions for instrument compatibility. Sterile. Color: clear.
Ordering information: All dishes include lids.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty