Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Griffin-Style Stainless Steel Beakers, Medegen
Supplier: Medegen Medical Products
84000 81200 80600 82000 81200 80125 80250
36311-320CS 1368.5 USD InStock
36311-320 14216-468 74360-008 74360-012 74360-013 74360-002 74360-010
Griffin-Style Stainless Steel Beakers, Medegen
Beakers
Type 304 stainless steel beakers feature a seamless construction for easy cleaning
Beakers are non-graduated and feature straight sides. These beakers feature a pour-spout and embossed fill lines on sides. No internal taper.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty