Would you like to log in again?
PEN MICRO TIP CONDUCTIVE
Supplier: ITW-CHEMTRONICS,INC
Conductive Ink Pen, Micro Tip^ Use this instrument on circuit boards to link components, make smooth jumpers, repair defective traces, and shield delicate electronics. Achieves maximum conductivity after 20 minutes, or after heat curing for 3-5 minutes at 121 to 149[degree]C.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
PEN MICRO TIP CONDUCTIVE 74555-008 CW2200MTP Case of 12 Retrieving
Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty