Would you like to log in again?
B.O.D. Bottles, Wheaton, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
227494-00 227494-03G 227494-03 227494-02 227494-00G 227494-02G 227494-99G 227494-01 227494-99 227494-01G
74830-048CS 1708.8 USD InStock
74830-048 80093-750 80093-748 74830-044 80093-744 74830-050 80093-746 16286-070 80093-740 80093-742
B.O.D. Bottles, Wheaton, WHEATON®, DWK Life Sciences
Bottles BOD Bottles
Ideal for incubating diluted samples of sewage, sewage effluents, polluted waters, and industrial wastes.
These 60 ml borosilicate glass bottles feature a shoulder radius that sweeps all air from inside the bottle during filling.
The bottles are designed with a permanently screened-on label and are available with either glass robotic stoppers or glass pennyhead stoppers. Bottles may be used with B.O.D. bottle rack. Bottle rack is rugged with plastic coating for corrosion resistance and quietness and holds twenty 60 ml B.O.D. bottles.
The interchangeable stoppers have a tapered bottom that also prevents air entrapment. Bottles have a flared mouth to form a water seal around the stopper that prevents air from being drawn inside during incubation.
Ordering information: For replacement bottles, contact your Avantor sales representative.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty