Would you like to log in again?
Glass Roller Culture Vessels, Tapered Neck, with Phenolic Caps, Wheaton®, DWK Life Sciences
Supplier: DWK Life Sciences
348253 348256 348252 348258
74830-056CS 557.5 USD InStock
74830-056 62404-621 62404-643 62404-600
Glass Roller Culture Vessels, Tapered Neck, with Phenolic Caps, Wheaton®, DWK Life Sciences
Bottles Roller Bottles
Vessels are made to exacting tolerances to provide higher cell yields.
  • Vessels and deep-skirted 38-415 screw caps are autoclavable
  • Free of glass imperfections
  • Neck area is tapered to facilitate cleaning
  • Caps have white styrene-butadiene rubber liner
Glass is specially selected for optical clarity and interior surface smoothness.
Certifications: Wheaton-33® low extractable borosilicate glass meets ASTM Type I Class A and USP Type I standards.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty