Would you like to log in again?
BOD Bottles, with Glass Robotic Stopper, 2 L, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences
227580
74830-128CS 215.51 USD InStock
74830-128
BOD Bottles, with Glass Robotic Stopper, 2 L, WHEATON®, DWK Life Sciences
Bottles BOD Bottles
Manufactured from WHEATON 33 low extractable borosilicate glass that conforms to USP Type I and also ASTM E 438 Type I Class A requirements and comes complete with a glass robotic stopper.
  • Recommended for long-term BOD and ultimate BOD analysis
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty