Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Staining Jar, 8-10 Slide Unit, Wheaton

Supplier: DWK Life Sciences

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

900620
74830-150CS 395.97 USD InStock
74830-150
Staining Jar, 8-10 Slide Unit, Wheaton
Jars Staining Jars
The WHEATON Staining Jar holds 8 single 3"x 1" (75 mm x 25 mm) slides vertically or 16 slides back-to-back

The jar's wide top is designed for convenience in transferring slides, making it especially suitable for staining slides that are inscribed on one end. The Jar and lid are manufactured from soda-lime glass, and the jar has an approximate inside bottom dimension of 47 mm L x 26 mm W x 85 mm H.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty