Would you like to log in again?
Nalgene® Validation Bottles, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
DS2127-0250 DS2127-0030 DS2127-2000
16172-153CS 202.56 USD InStock
16172-153 74910-034 74910-032
Nalgene® Validation Bottles, Thermo Scientific
Bottles Carboys
Ideal for QA lot sample storage of pharmaceuticals.
  • Tough, nontoxic containers are autoclavable for sterile applications
Bottles allow convenient material compatibility validation when scaling up production to larger containers. Constructed of clear polycarbonate and supplied with white polypropylene closures with TPE gasket. Bottles feature thick walls and wide mouths. Transparent design allows for visual inspection of contents.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty