Would you like to log in again?
SP Bel-Art Write-On™ Bottles, HDPE, Bel-Art Products, a part of SP
Supplier: Bel-Art
SCIENCEWARE®
F10660-0500 F10662-1000 F10660-1000 F10662-0500
74950-035PK 39.64 USD InStock
74950-035 74950-033 74950-031 74950-029
SP Bel-Art Write-On™ Bottles, HDPE, Bel-Art Products, a part of SP
Bottles Plastic Bottles
Molded from rugged high-density polyethylene in 500 and 1000 ml (16.9 and 33.8 oz.) sizes, these bottles are available in narrow (28 mm) or wide mouth (53 mm) formats, and have leakproof polypropylene caps.
  • Rugged HDPE construction
  • Leakproof polypropylene caps
  • Ideal for use with both liquids or dry materials
Screened labels include a name identification area and a large detail area for contents or safety information, as well as a defined space for date information. Ideal for storing and transporting liquids or dry material.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty