Would you like to log in again?
SP Bel-Art Aquet® Detergent for Glassware and Plastics, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a part of SP
Aquet®
F17094-0020 F17094-0050 F17094-0030
75783-970EA 96.08 USD InStock
75783-970 74950-097 54848-004
SP Bel-Art Aquet® Detergent for Glassware and Plastics, Bel-Art Products, a part of SP
Cleaning Agents Detergents and Surface Cleansers
Safe for glassware, plastics, and surfaces, this phosphate-free, nonionic surfactant will not etch or mar precision glassware.
  • Convenient, pre-portioned pouches make one gallon of 1% solution
  • 3.8 L (1 gal.) bottle comes with 473 ml (16 oz.) working solution dispenser bottle
  • 20 L (5 gal.) Cubitainer™ is available for frequent users
  • Not for use in automatic dishwashers
It rinses quickly and dries without wiping.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty