Would you like to log in again?
SP Bel-Art Cryo Tower Storage Systems, Bel-Art Products, a part of SP
Supplier: Bel-Art
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Cryo-Safe™ SP Bel-Art
F18853-0004 F18853-0008
74950-134EA 173.34 USD InStock
74950-134 74950-132
SP Bel-Art Cryo Tower Storage Systems, Bel-Art Products, a part of SP
Boxes Cryoboxes
These polycarbonate storage towers are ideal for benchtop or incubator use, transportation, freezer, and general-purpose storage.
  • Polycarbonate construction
  • Small bench space footprint
  • Autoclavable
A 15.2x15.2 cm (6x6") footprint saves valuable bench space.
Available in 4- and 8-level configurations, both towers accommodate Cryo-Safe™ Vial Storage Box 47751-740. Autoclavable at 121°C (250°F).
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty