Would you like to log in again?
SP Bel-Art Centrifuge Tube Refrigerator Racks, 50 ml, Bel-Art Products, a part of SP
Supplier: Bel-Art
F18905-0050 F18905-0051
74950-140EA 37.83 USD InStock
74950-140 74950-138
SP Bel-Art Centrifuge Tube Refrigerator Racks, 50 ml, Bel-Art Products, a part of SP
Racks Tube Racks
This all-polypropylene rack provides efficient and extended cooling of twenty-eight 50 mL centrifuge tubes in a chilled bath.
  • Full polypropylene construction
  • Flat perimeter rim
  • Autoclavable
  • Optional polypropylene cover protects tubes during transport or storage
It holds approximately 2L (67oz.) of ice and water and allows the tubes to rest on the bottom for maximum stability and exposure to the bath.
A flat perimeter rim provides space for labeling tape. Steam autoclavable at 121°C (250°F) and stackable with identical racks when full or empty. An optional polypropylene cover is available to protect tubes during transport or storage while maintaining stackability.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty