Would you like to log in again?
SPOON MINI SAMPLER .25ML
Supplier: Bel-Art Products, a Part of SP
Mini Sampler^ Size: 0.25mL. Ideal for retrieving small, specific volume samples of powders or similar materials. Use a separate spoon for each material sampled to avoid cross contamination. Molded of white high impact polystyrene. 25 per bag, 10 bags per case.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SPOON MINI SAMPLER .25ML 74950-174 F36721-0025 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty