Would you like to log in again?
SP Bel-Art Economy Sampling Scoops, Bel-Art Products, a part of SP
Supplier: Bel-Art
Sterileware®
F36933-0022 F36933-0004 F36933-0001 F36933-0023 F36933-0021 F36933-0024 F36933-0002 F36933-0003
74950-213PK 29.82 USD InStock
74950-213 74950-203 74950-211 74950-201 74950-209 74950-207 74950-215 74950-205
SP Bel-Art Economy Sampling Scoops, Bel-Art Products, a part of SP
Scoops
Designed for accurate measurement of small quantities of dry or liquid substances.
  • One-piece design
  • Polystyrene construction
  • Durable
  • Lightweight
  • Flat-bottom shape
One-piece scoops are molded from durable, FDA-grade, high-impact polystyrene plastic. Leveling the sample to the flat top edge provides accurate and repeatable volume measurements. Flat-bottom shape and light weight allow use as a weighing boat.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty