Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Polygon Spinbar Magnetic Stirring Bars
Supplier: VWR
VWR® Spinbar®
F37122-0070 F37122-0025 F37122-0040 F37122-0045 F37122-0060 F37122-0050 F37122-0012 F37122-0020 F37122-0035
74950-288EA 8.95 USD InStock
74950-288 74950-300 74950-286 74950-298 74950-284 74950-296 74950-294 74950-292 74950-290
VWR® Polygon Spinbar Magnetic Stirring Bars
Stirrers Magnetic Stirring Bars
These VWR® stirbars have polygon multifaceted surfaces that add turbulence relative to similar smooth size cylindrical bars
  • Center molded pivot ring minimizes the contact area between the bar and the vessel; reduces friction and chattering
  • Marring of plastic containers is diminished
  • Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon® brand PTFE Coating – Superior Chemical Compatibility
  • Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
  • Autoclave safe
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty