Would you like to log in again?
SP Bel-Art Dry-Keeper™ Small, Stacking Desiccator Cabinet, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a part of SP
Dry-Keeper™
H42053-0000
74950-336EA 255.84 USD InStock
74950-336
SP Bel-Art Dry-Keeper™ Small, Stacking Desiccator Cabinet, Bel-Art Products, a part of SP
Desiccators Desiccator Cabinets
Small, clear polystyrene desiccator cabinet safely stacks up to 5 units high for maximum use of dry storage space.
  • Polystyrene construction
  • Door disappears beneath cabinet when open
  • Non-electric
  • Stackable design
Pull-down door slides out of the way into a compartment located underneath the cabinet.
Made of high-impact polystyrene. Completely transparent for full visibility from any side. Foam rubber gasket forms seal when door is latched with two side roller latches. When opened, door disappears underneath cabinet. May be stacked five high to save bench space.
Ordering information: Ready to use jar of silica gel is included.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty